missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Renin-2 Protein
A DNA sequence encoding the Mus musculus (Mouse) Renin-2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8886-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_112470.2 | |
Renin-2 Protein | |
Research Use Only | |
Ren2 | |
Recombinant Protein | |
E. coli | |
SSLTDLISPV VLTNYLNSQY YGEIGIGTPP QTFKVIFDTG SANLWVPSTK CSRLYLACGI HSLYESSDSS SYMENGDDFT IHYGSGRVKG FLSQDSVTVG GITVTQTFGE VTELPLIPFM LAQFDGVLGM GFPAQAVGGV TPVFDHILSQ GVLKEKVFSV YYNRGPHLLG GEVVLGGSDP EHYQGDFHYV SLSKTDSWQI TMKGVSVGSS TLLCEEGCEV VVDTGSSFIS APTSSLKLIM QALGAKEKRL HEYVVSCSQV PTLPDISFNL GGRAYTLSST DYVLQYPN | |
Greater than 90% as determined by SDS-PAGE. |
19702 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |