missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Proto-oncogene Wnt-3 Protein
A DNA sequence encoding the Mus musculus (Mouse) Proto-oncogene Wnt-3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9467-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_033547.1 | |
Proto-oncogene Wnt-3 Protein | |
Research Use Only | |
Wnt3 | |
Recombinant Protein | |
Yeast | |
GYPIWWSLAL GQQYTSLASQ PLLCGSIPGL VPKQLRFCRN YIEIMPSVAE GVKLGIQECQ HQFRGRRWNC TTIDDSLAIF GPVLDKATRE SAFVHAIASA GVAFAVTRSC AEGTSTICGC DSHHKGPPGE GWKWGGCSED ADFGVLVSRE FADARENRPD ARSAMNKHNN EAGRTTILDH MHLKCKCHGL SGSCEVKTCW WAQPDFRAIG DFLKDKYDSA SEMVVEKHRE SRGWVETLRA KYALFKPPTE RDLVYYENSP NFCEPNPETG SFGTRDRTCN VTSHGIDGCD LLCCGRGHNT RTEKRKEKCH CVF | |
Greater than 90% as determined by SDS-PAGE. |
22415 | |
50 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |