missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse PKM2 Protein

Product Code. p-7148635
Change view
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15915288 2 μg 2µg
15995278 10 μg 10µg
15905288 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15915288 Supplier enQuireBio™ Supplier No. QP108252ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the PKM2 was constructed and used to recombinantly synthesize the protein.

Specifications

Name PKM2 Protein
Quantity 2 μg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Biological Activity Specific activity: > 500 units/mg. One unit will convert 1 pmole of phospho(enol)pyruvate to pyruvate per minute at pH 7.5 at 37C.
Product Type Recombinant Protein
Cross Reactivity Mouse
Species E. coli
Protein Tag His
Sequence MGSSHHHHHH SSGLVPRGSH MGSMPKPHSE AGTAFIQTQQ LHAAMADTFL EHMCRLDIDS APITARNTGI ICTIGPASRS VEMLKEMIKS GMNVARLNFS HGTHEYHAET IKNVREATES FASDPILYRP VAVALDTKGP EIRTGLIKGS GTAEVELKKG ATLKITLDNA YMEKCDENIL WLDYKNICKV VEVGSKIYVD DGLISLQVKE KGADFLVTEV ENGGSLGSKK GVNLPGAAVD LPAVSEKDIQ DLKFGVEQDV DMVFASFIRK AADVHEVRKV LGEKGKNIKI ISKIENHEGV RRFDEILEAS DGIMVARGDL GIEIPAEKVF LAQKMMIGRC NRAGKPVICA TQMLESMIKK PRPTRAEGSD VANAVLDGAD CIMLSGETAK GDYPLEAVRM QHLIAREAEA AIYHLQLFEE LRRLAPITSD PTEAAAVGAV EASFKCCSGA IIVLTKSGRS AHQVARYRPR APIIAVTRNP QTARQAHLYR GIFPVLCKDA VLNAWAEDVD LRVNLAMDVG KARGFFKKGD VVIVLTGWRP GSGFTNTMRV VPVP
Buffer PKM2 protein solution (0.5 mg/ml) containing 20mM Tris-HCl (pH 8.5), 0.2M NaCl, 1mM DTT and 30% glycerol.
Purity or Quality Grade Greater than 85% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.