missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse p53 / TP53 Protein
A DNA sequence encoding the Mus musculus (Mouse) p53 / TP53, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6823-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001120705.1 | |
p53 / TP53 Protein | |
Research Use Only | |
TP53 | |
Recombinant Protein | |
E. coli | |
MEESQSDISL ELPLSQETFS GLWKLLPPED ILPSPHCMDD LLLPQDVEEF FEGPSEALRV SGAPAAQDPV TETPGPVAPA PATPWPLSSF VPSQKTYQGN YGFHLGFLQS GTAKSVMCTY SPPLNKLFCQ LAKTCPVQLW VSATPPAGSR VRAMAIYKKS QHMTEVVRRC PHHERCSDGD GLAPPQHLIR VEGNLYPEYL EDRQTFRHSV VVPYEPPEAG SEYTTIHYKY MCNSSCMGGM NRRPILTIIT LEDSSGNLLG RDSFEVRVCA CPGRDRRTEE ENFRKKEVLC PELPPGSAKR ALPTCTSASP PQKKKPLDGE YFTLKIRGRK RFEMFRELNE ALELKDAHAT EESGDSRAHS SYLKTKKGQS TSRHKKTMVK KVGPDSD | |
Greater than 90% as determined by SDS-PAGE. |
22059 | |
500 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |