missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse MCL1 homolog Protein
A DNA sequence encoding the Mus musculus (Mouse) MCL1 homolog, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6348-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_032588.1 | |
MCL1 homolog Protein | |
Research Use Only | |
Mcl1 | |
Recombinant Protein | |
E. coli | |
MFGLRRNAVI GLNLYCGGAS LGAGGGSPAG ARLVAEEAKA RREGGGEAAL LPGARVVARP PPVGAEDPDV TASAERRLHK SPGLLAVPPE EMAASAAAAI VSPEEELDGC EPEAIGKRPA VLPLLERVSE AAKSSGADGS LPSTPPPPEE EEDDLYRQSL EIISRYLREQ ATGSKDSKPL GEAGAAGRRA LETLRRVGDG VQRNHETAFQ GMLRKLDIKN EGDVKSFSRV MVHVFKDGVT NWGRIVTLIS FGAFVAKHLK SVNQESFIEP LAETITDVLV RTKRDWLVKQ RGWDGFVEFF HVQDLEGG | |
Greater than 90% as determined by SDS-PAGE. |
17210 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |