missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL1F10 / IL-38 Protein
A DNA sequence encoding the Mus musculus (Mouse) IL1F10 / IL-38, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7885-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_694717.1 | |
IL1F10 / IL-38 Protein | |
Research Use Only | |
Il1f10 | |
Recombinant Protein | |
Yeast | |
MCSLPMARYY IIKDAHQKAL YTRNGQLLLG DPDSDNYSPE KVCILPNRGL DRSKVPIFLG MQGGSCCLAC VKTREGPLLQ LEDVNIEDLY KGGEQTTRFT FFQRSLGSAF RLEAAACPGW FLCGPAEPQQ PVQLTKESEP STHTEFYFEM SR | |
Greater than 80% as determined by SDS-PAGE. |
215274 | |
1 mg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |