missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse IL-23 (HEK293-expressed) Protein
Click to view available options
Quantity:
10 μg
250 μg
50 μg
Unit Size:
10µg
250µg
50µg
Description
Measured by its ability to induce IL-17 secretion by mouse splenocytes. Aggarwal, S. et al. (2003) J. Biol. Chem. 278:1910. The ED50 for this effect is 0.050-0.500 ng/mL.
Specifications
Specifications
| Accession Number | P43432.1 (p40) & Q9EQ14.1 (p19) |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Molecular Weight (g/mol) | MolecularWeight-observed: 60-75 kDa, under reducing conditions., MolecularWeight-theroretical: 57 kDa |
| Quantity | 250 μg |
| Source | Human embryonic kidney cell, HEK293-derived mouse IL-23 protein Mouse IL-12p40 (Met22-Ser335) Accession # P43432.1 GSGSSRGGSGSGGSGGGGSKL Mouse IL-23p19 (Leu20-Ala196) Accession # Q9EQ14.1 (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Reconstitution | Reconstitute at 100-500 μg/mL in PBS. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction