missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Egl nine homolog 3 Protein

A DNA sequence encoding the Mus musculus (Mouse) Egl nine homolog 3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8813-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16046621

  • 762.00 € / 100µg

Estimated Shipment: 16-11-2022
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_082409.2
Egl nine homolog 3 Protein
100 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
112407
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Egln3
Recombinant Protein
E. coli
PLGHIMRLDL EKIALEYIVP CLHEVGFCYL DNFLGEVVGD CVLERVKQLH YNGALRDGQL AGPRAGVSKR HLRGDQITWI GGNEEGCEAI NFLLSLIDRL VLYCGSRLGK YYVKERSKAM VACYPGNGTG YVRHVDNPNG DGRCITCIYY LNKNWDAKLH GGVLRIFPEG KSFVADVEPI FDRLLFFWSD RRNPHEVQPS YATRYAMTVW YFDAEERAEA KKKFRNLTRK TESALAKD
Videos
SDS
Documents
Certificates
Promotions

Promotions