missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Cystathionine beta-synthase Protein
A DNA sequence encoding the Mus musculus (Mouse) Cystathionine beta-synthase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7956-ye-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001258282.1 | |
Cystathionine beta-synthase Protein | |
Research Use Only | |
CBS | |
Recombinant Protein | |
Yeast | |
PSGTSQCEDG SAGGFQHLDM HSEKRQLEKG PSGDKDRVWI RPDTPSRCTW QLGRAMADSP HYHTVLTKSP KILPDILRKI GNTPMVRINK ISKNAGLKCE LLAKCEFFNA GGSVKDRISL RMIEDAERAG NLKPGDTIIE PTSGNTGIGL ALAAAVKGYR CIIVMPEKMS MEKVDVLRAL GAEIVRTPTN ARFDSPESHV GVAWRLKNEI PNSHILDQYR NASNPLAHYD DTAEEILQQC DGKLDMLVAS AGTGGTITGI ARKLKEKCPG CKIIGVDPEG SILAEPEELN QTEQTAYEVE GIGYDFIPTV LDRAVVDKWF KSNDEDSFAF ARMLIAQEGL LCGGSSGSAM AVAVKAAREL QEGQRCVVIL PDSVRNYMSK FLSDKWMLQK GFMKEELSVK RPWWWRLRVQ ELSLSAPLTV LPTVTCEDTI AILREKGFDQ APVVNESGAI LGMVTLGNML SSLLAGKVRP SDEVCKVLYK QFKPIHLTDT LGTLSHILEM DHFALVVHEQ IQSRDQAWSG VVGGPTDCSN GMSSKQQMVF GVVTAIDLLN FVAAREQTQT | |
Greater than 90% as determined by SDS-PAGE. |
12411 | |
500 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |