missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse CD42b/GPIb alpha Protein
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
The Recombinant Mouse CD42b/GPIb alpha Protein is derived from CHO. The Recombinant Mouse CD42b/GPIb alpha Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Accession Number | O35930 |
| For Use With (Application) | Bioactivity |
| Gene ID (Entrez) | 2811 |
| Molecular Weight (g/mol) | Observed M.W.: 108-150kDa, reducing conditions, Theoretical M.W.: 66kDa |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived mouse CD42b/GPIb alpha protein Gln17-Pro612,with a C-terminal 6-His tag Accession &Num; O35930 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1μg of the protein by the LAL method. |
| Gene Alias | Antigen CD42b-alpha, BP1BA, BSS, CD42b, CD42b antigen, CD42b-alpha, glycoprotein Ib (platelet), alpha polypeptide, Glycoprotein Ibalpha, GP1B, GP1BA, GPIb alpha, GP-Ib alpha, GPIbA, GPIb-alpha, MGC34595, platelet glycoprotein Ib alpha chain, platelet membrane glycoprotein 1b-alpha subunit |
| Common Name | CD42b/GPIb alpha |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction