missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Mouse Caspase-1 Protein

Product Code. 18066778 Shop All Bio Techne Products
Click to view available options
Quantity:
100 Each
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 18066778

Brand: Novus Biologicals™ NBP199607100EACH

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified and high bioactivity. Generating reliable and reproducible results.

Recombinant protein for Mouse Caspase-1 The Recombinant Mouse Caspase-1 Protein is derived from E. coli. The Recombinant Mouse Caspase-1 Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Lyoph
For Use With (Application) Bioactivity
Formulation Semi-Dry
Gene ID (Entrez) 834
Quantity 100 Each
Source E. coli
Storage Requirements Store at -20 to -80C. Avoid freeze-thaw cycles.
Gene Alias CASP-1, caspase 1, apoptosis-related cysteine peptidase (interleukin 1, beta, caspase 1, apoptosis-related cysteine protease (interleukin 1, beta, caspase-1, EC 3.4.22, EC 3.4.22.36, ICEP45, IL-1 beta-converting enzyme, IL-1BC, IL1BC CASP1 nirs variant 1, IL1BCE, IL1B-convertase, interleukin 1-B converting enzyme, interleukin 1-beta convertase, Interleukin-1 beta convertase, Interleukin-1 beta-converting enzyme, p45
Gene Symbol CASP1
Reconstitution Reconstitute to 1 unit per μL in PBS containing 15% glycerol.
Buffer Semi-Dry
Purity or Quality Grade SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.