missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Carboxylesterase 1C Protein
A DNA sequence encoding the Mus musculus (Mouse) Carboxylesterase 1C, was expressed in the hosts and tags indicated.
299.00 € - 3175.00 €
Specifications
Accession Number | NP_031980.2 |
---|---|
Gene ID (Entrez) | 13884 |
Name | Carboxylesterase 1C Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16047620
|
enQuireBio™
QP7188-YE-100UG |
100 μg |
960.00 €
100µg |
Estimated Shipment: 08-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16087620
|
enQuireBio™
QP7188-YE-500UG |
500 μg |
2650.00 €
500µg |
Estimated Shipment: 08-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16057620
|
enQuireBio™
QP7188-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16077620
|
enQuireBio™
QP7188-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16097620
|
enQuireBio™
QP7188-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16067620
|
enQuireBio™
QP7188-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_031980.2 | |
Carboxylesterase 1C Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
13884 | |
Research Use Only | |
Ces1c | |
Recombinant Protein | |
Yeast | |
HSLLPPVVDT TQGKVLGKYI SLEGFEQPVA VFLGVPFAKP PLGSLRFAPP QPAEPWSFVK NATSYPPMCS QDAGWAKILS DMFSTEKEIL PLKISEDCLY LNIYSPADLT KSSQLPVMVW IHGGGLVIGG ASPYNGLALS AHENVVVVTI QYRLGIWGLF STGDEHSPGN WAHLDQLAAL RWVQDNIANF GGNPDSVTIF GESSGGISVS VLVLSPLGKD LFHRAISESG VVINTNVGKK NIQAVNEIIA TLSQCNDTSS AAMVQCLRQK TESELLEISG KLVQYNISLS TMIDGVVLPK APEEILAEKS FNTVPYIVGF NKQEFGWIIP MMLQNLLPEG KMNEETASLL LRRFHSELNI SESMIPAVIE QYLRGVDDPA KKSELILDMF GDIFFGIPAV LLSRSLRDAG VSTYMYEFRY RPSFVSDKRP QTVEGDHGDE IFFVFGAPLL KEGASEEETN LSKMVMKFWA NFARNGNPNG EGLPHWPEYD EQEGYLQIGA TTQQAQRLKA EEVAFWTELL AKNPPETDPT EH | |
Greater than 90% as determined by SDS-PAGE. |