missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Macaque GM-CSF Protein

Product Code. 15963878
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 μg
250 μg
Unit Size:
100µg
10µg
250µg
2µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15963878 2 μg 2µg
15943878 10 μg 10µg
15933878 100 μg 100µg
15953878 250 μg 250µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15963878 Supplier enQuireBio™ Supplier No. QP10445ec2ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Rhesus Macaque GM-CSF, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001028121.1
Gene ID (Entrez) 574371
Name GM-CSF Protein
Quantity 2 μg
Regulatory Status Research Use Only
Endotoxin Concentration Less than 1.0 EU/ug as determined by LAL method.
Gene Symbol GM-CSF
Protein Length Partial (see sequence information for more details).
Biological Activity The ED50 as determined by a cell proliferation assay using human TF-1 cells is less than 0.1 ng/ml, corresponding to a specific activity of > 1.0 M 107 IU/mg.
Product Type Recombinant Protein
Cross Reactivity Macaque
Species E. coli
Protein Tag Untagged
Sequence APARSPSPGT QPWEHVNAIQ EARRLLNLSR DTAAEMNKTV EVVSEMFDLQ EPSCLQTRLE LYKQGLQGSL TKLKGPLTMM ASHYKQHCPP TPETSCATQI ITFQSFKENL KDFLLVIPFD CWEPVQE
Buffer 0.2 um filtered PBS, pH 7.4, lyophilized
Purity or Quality Grade Greater than 98% as determined by SDS-PAGE and HPLC.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.