missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Zymogen granule protein 16 homolog B Protein

A DNA sequence encoding the Homo sapiens (Human) Zymogen granule protein 16 homolog B, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7869-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16041271

  • 762.00 € / 100µg

Estimated Shipment: 18-11-2022
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_660295.2
Zymogen granule protein 16 homolog B Protein
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
124220
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ZG16B
Recombinant Protein
E. coli
GKMYGPGGGK YFSTTEDYDH EITGLRVSVG LLLVKSVQVK LGDSWDVKLG ALGGNTQEVT LQPGEYITKV FVAFQAFLRG MVMYTSKDRY FYFGKLDGQI SSAYPSQEGQ VLVGIYGQYQ LLGIKSIGFE WNYPLEEPTT EPPVNLTYSA NSPVGR
Videos
SDS
Documents
Certificates
Promotions

Promotions