missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human VEGF-C Protein

A DNA sequence encoding the Homo sapiens (Human) VEGF-C, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8577-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16045381

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_005420.1
VEGF-C Protein
Research Use Only
VEGFC
Recombinant Protein
E. coli
AHYNTEILKS IDNEWRKTQC MPREVCIDVG KEFGVATNTF FKPPCVSVYR CGGCCNSEGL QCMNTSTSYL SKTLFEITVP LSQGPKPVTI SFANHTSCRC MSKLDVYRQV HSIIRR
Greater than 90% as determined by SDS-PAGE.
7424
200 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions