missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UBL3 Protein
Specifications
Specifications
| Name | UBL3 Protein |
| Quantity | 5 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGSSHHHHHH SSGLVPRGSH MSSNVPADMI NLRLILVSGK TKEFLFSPND SASDIAKHVY DNWPMDWEEE QVSSPNILRL IYQGRFLHGN VTLGALKLPF GKTTVMHLVA RETLPEPNSQ GQRNREKTGE SNCC |
| Buffer | UBL3 protein solution (1 mg/1ml) containing 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 0.1M NaCl and 2mM DTT. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?