missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Trypsin 1/PRSS1 Protein

Product Code. 18118160
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18118160 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18118160 Supplier R&D Systems™ Supplier No. 3848SE010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to cleave the fluorogenic peptide substrate, Mca-RPKPVE-Nval-WRK(Dnp)-NH2 (Catalog # ES002 ). The specific activity is >3,000 pmol/min/μg, as measured under the described conditions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_002760
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in HCl and NaCl.
Gene ID (Entrez) 5644
Molecular Weight (g/mol) M.W. (Observed): 32 kDa and 36 kDa, reducing conditions; M.W. (theoretical): 26 kDa
Name Trypsin 1/PRSS1
Quantity 10 μg
Source Mouse myeloma cell line, NS0-derived human Trypsin 1/PRSS1 protein Ala16-Ser247, with a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Structural Form Pro form
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol PRSS1
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein Trypsin 1/PRSS1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.