missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Transmembrane protein 14B Protein
A DNA sequence encoding the Homo sapiens (Human) Transmembrane protein 14B, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6804-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001121183.1 | |
Transmembrane protein 14B Protein | |
Research Use Only | |
TMEM14B | |
Recombinant Protein | |
E. coli | |
MEKPLFPLVP LHWFGFGYTA LVVSGGIVGY VKTGSVPSLA AGLLFGSLAG LGAYQLYQDP RNVWGFLAAT SVTFVGVMGM RSYYYGKFMP VGLIAGASLL MAAKVGVRML MTSD | |
Greater than 90% as determined by SDS-PAGE. |
81853 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |