missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Transcriptional enhancer factor TEF-1 Protein

A DNA sequence encoding the Homo sapiens (Human) Transcriptional enhancer factor TEF-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9426-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16098281

  • 645.00 € / 100µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_068780.2
Transcriptional enhancer factor TEF-1 Protein
Research Use Only
TEAD1
Recombinant Protein
Yeast
MEPSSWSGSE SPAENMERMS DSADKPIDND AEGVWSPDIE QSFQEALAIY PPCGRRKIIL SDEGKMYGRN ELIARYIKLR TGKTRTRKQV SSHIQVLARR KSRDFHSKLK DQTAKDKALQ HMAAMSSAQI VSATAIHNKL GLPGIPRPTF PGAPGFWPGM IQTGQPGSSQ DVKPFVQQAY PIQPAVTAPI PGFEPASAPA PSVPAWQGRS IGTTKLRLVE FSAFLEQQRD PDSYNKHLFV HIGHANHSYS DPLLESVDIR QIYDKFPEKK GGLKELFGKG PQNAFFLVKF WADLNCNIQD DAGAFYGVTS QYESSENMTV TCSTKVCSFG KQVVEKVETE YARFENGRFV YRINRSPMCE YMINFIHKLK HLPEKYMMNS VLENFTILLV VTNRDTQETL LCMACVFEVS NSEHGAQHHI YRLVKD
Greater than 90% as determined by SDS-PAGE.
7003
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions