missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human T-cell receptor beta-1 chain C region Protein
A DNA sequence encoding the Homo sapiens (Human) T-cell receptor beta-1 chain C region, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9445-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
T-cell receptor beta-1 chain C region Protein | |
Research Use Only | |
TRBC1 | |
Recombinant Protein | |
Yeast | |
EDLNKVFPPE VAVFEPSEAE ISHTQKATLV CLATGFFPDH VELSWWVNGK EVHSGVSTDP QPLKEQPALN DSRYCLSSRL RVSATFWQNP RNHFRCQVQF YGLSENDEWT QDRAKPVTQI VSAEAWGRAD CGFTSVSYQQ GVLSATILYE ILLGKATLYA VLVSALVLMA MVKRKDF | |
Greater than 90% as determined by SDS-PAGE. |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |