missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Syntaxin-11 Protein
A DNA sequence encoding the Homo sapiens (Human) Syntaxin-11, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6742-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003755.2 | |
Syntaxin-11 Protein | |
Research Use Only | |
STX11 | |
Recombinant Protein | |
E. coli | |
MKDRLAELLD LSKQYDQQFP DGDDEFDSPH EDIVFETDHI LESLYRDIRD IQDENQLLVA DVKRLGKQNA RFLTSMRRLS SIKRDTNSIA KAIKARGEVI HCKLRAMKEL SEAAEAQHGP HSAVARISRA QYNALTLTFQ RAMHDYNQAE MKQRDNCKIR IQRQLEIMGK EVSGDQIEDM FEQGKWDVFS ENLLADVKGA RAALNEIESR HRELLRLESR IRDVHELFLQ MAVLVEKQAD TLNVIELNVQ KTVDYTGQAK AQVRKAVQYE EKNPCRTLCC FCCPCLK | |
Greater than 80% as determined by SDS-PAGE. |
8676 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |