missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human STAT3 Protein
A DNA sequence encoding the Homo sapiens (Human) STAT3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9416-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003141.2 | |
STAT3 Protein | |
Research Use Only | |
STAT3 | |
Recombinant Protein | |
Yeast | |
MAQWNQLQQL DTRYLEQLHQ LYSDSFPMEL RQFLAPWIES QDWAYAASKE SHATLVFHNL LGEIDQQYSR FLQESNVLYQ HNLRRIKQFL QSRYLEKPME IARIVARCLW EESRLLQTAA TAAQQGGQAN HPTAAVVTEK QQMLEQHLQD VRKRVQDLEQ KMKVVENLQD DFDFNYKTLK SQGDMQDLNG NNQSVTRQKM QQLEQMLTAL DQMRRSIVSE LAGLLSAMEY VQKTLTDEEL ADWKRRQQIA CIGGPPNICL DRLENWITSL AESQLQTRQQ IKKLEELQQK VSYKGDPIVQ HRPMLEERIV ELFRNLMKSA FVVERQPCMP MHPDRPLVIK TGVQFTTKVR LLVKFPELNY QLKIKVCIDK DSGDVAALRG SRKFNILGTN TKVMNMEESN NGSLSAEFKH LTLREQRCGN GGRANCDASL IVTEELHLIT FETEVYHQGL KIDLETHSLP VVVISNICQM PNAWASILWY NMLTNNPKNV NFFTKPPIGT WDQVAEVLSW QFSSTTKRGL SIEQLTTLAE KLLGPGVNYS GCQITWAKFC KENMAGKGFS FWVWLDNIID LVKKYILALW NEGYIMGFIS KERERAILST KPPGTFLLRF SESSKEGGVT FTWVEKDISG KTQIQSVEPY TKQQLNNMSF AEIIMGYKIM DATNILVSPL VYLYPDIPKE EAFGKYCRPE SQEHPEADPG SAAPYLKTKF ICVTPTTCSN TIDLPMSPRT LDSLMQFGNN GEGAEPSAGG QFESLTFDME LTSECATSPM | |
Greater than 90% as determined by SDS-PAGE. |
6774 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |