missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Spermidine synthase Protein
A DNA sequence encoding the Homo sapiens (Human) Spermidine synthase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8349-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003123.2 | |
Spermidine synthase Protein | |
Research Use Only | |
SRM | |
Recombinant Protein | |
E. coli | |
REGWFRETCS LWPGQALSLQ VEQLLHHRRS RYQDILVFRS KTYGNVLVLD GVIQCTERDE FSYQEMIANL PLCSHPNPRK VLIIGGGDGG VLREVVKHPS VESVVQCEID EDVIQVSKKF LPGMAIGYSS SKLTLHVGDG FEFMKQNQDA FDVIITDSSD PMGPAESLFK ESYYQLMKTA LKEDGVLCCQ GECQWLHLDL IKEMRQFCQS LFPVVAYAYC TIPTYPSGQI GFMLCSKNPS TNFQEPVQPL TQQQVAQMQL KYYNSDVHRA AFVLPEFARK ALNDVS | |
Greater than 90% as determined by SDS-PAGE. |
6723 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |