missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Small nuclear ribonucleoprotein Sm D2 Protein

A DNA sequence encoding the Homo sapiens (Human) Small nuclear ribonucleoprotein Sm D2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8419-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16044511

  • 518.00 € / 100µg

Estimated Shipment: 16-11-2022
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_004588.1
Small nuclear ribonucleoprotein Sm D2 Protein
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
6633
Greater than 90% as determined by SDS-PAGE.
Research Use Only
SNRPD2
Recombinant Protein
E. coli
MSLLNKPKSE MTPEELQKRE EEEFNTGPLS VLTQSVKNNT QVLINCRNNK KLLGRVKAFD RHCNMVLENV KEMWTEVPKS GKGKKKSKPV NKDRYISKMF LRGDSVIVVL RNPLIAGK
Videos
SDS
Documents
Certificates
Promotions

Promotions