missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Sjoegren syndrome nuclear autoantigen 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Sjoegren syndrome nuclear autoantigen 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6732-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003722.2 | |
Sjoegren syndrome nuclear autoantigen 1 Protein | |
Research Use Only | |
SSNA1 | |
Recombinant Protein | |
E. coli | |
TQQGAALQNY NNELVKCIEE LCQKREELCR QIQEEEDEKQ RLQNEVRQLT EKLARVNENL ARKIASRNEF DRTIAETEAA YLKILESSQT LLSVLKREAG NLTKATAPDQ KSSGGRDS | |
Greater than 80% as determined by SDS-PAGE. |
8636 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |