Learn More
R&D Systems™ Recombinant Human Sirtuin 3/SIRT3 Protein
Description
The Recombinant Human Sirtuin 3/SIRT3 Protein is derived from E. coli. The Recombinant Human Sirtuin 3/SIRT3 Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in Tris, NaCl, Brij-35 and Glycerol. |
| Gene ID (Entrez) | 23410 |
| Molecular Weight (g/mol) | 34 kDa |
| Name | Sirtuin 3/SIRT3 |
| Quantity | 50μg |
| Source | E. coli-derived human Sirtuin 3/SIRT3 protein Ser101-Lys399,with an N-terminal Met and 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain at 5 μg per lane. |