missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Sialic acid-binding Ig-like lectin 15 Protein
A DNA sequence encoding the Homo sapiens (Human) Sialic acid-binding Ig-like lectin 15, was expressed in the hosts and tags indicated.
605.00 € - 1690.00 €
Specifications
Accession Number | NP_998767.1 |
---|---|
Gene ID (Entrez) | 284266 |
Name | Sialic acid-binding Ig-like lectin 15 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16000721
|
enQuireBio™
QP7774-MA-20UG |
20 μg |
605.00 €
20µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16090711
|
enQuireBio™
QP7774-MA-100UG |
100 μg |
1690.00 €
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16010721
|
enQuireBio™
QP7774-MA-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_998767.1 | |
Sialic acid-binding Ig-like lectin 15 Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
284266 | |
Research Use Only | |
SIGLEC15 | |
Recombinant Protein | |
Mammal | |
FVRTKIDTTE NLLNTEVHSS PAQRWSMQVP PEVSAEAGDA AVLPCTFTHP HRHYDGPLTA IWRAGEPYAG PQVFRCAAAR GSELCQTALS LHGRFRLLGN PRRNDLSLRV ERLALADDRR YFCRVEFAGD VHDRYESRHG VRLHVTAAPR IVNISVLPSP AHAFRALCTA EGEPPPALAW SGPALGNSLA AVRSPREGHG HLVTAELPAL THDGRYTCTA ANSLGRSEAS VYLFRFHGAS GAST | |
Greater than 90% as determined by SDS-PAGE. |