missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Sex comb on midleg-like protein 1
A DNA sequence encoding the Homo sapiens (Human) Sex comb on midleg-like protein 4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7903-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001273337.1 | |
Sex comb on midleg-like protein 1 | |
Research Use Only | |
SCML4 | |
Recombinant Protein | |
E. coli | |
MPCQRTAWIG CDRQRTPPFH WREIKSRVLM TPLALSPPRS TPEPDLSSIP QDAATVPSLA APQALTVCLY INKQANAGPY LERKKVQQLP EHFGPERPSA VLQQAVQACI DCAHQQKLVF SLVKQGYGGE MVSVSASFDG KQHLRSLPVV NSIGYVLRFL AKLCRSLLCD DLFSHQPFPR GCSASEKVQE KEEGRMESVK TVTTEEYLVN PVGMNRYSVD TSASTFNHRG SLHPSSSLYC KRQNSGDSHL GGGPAATAGG PRTSPMSSGG PSAPGLRPPA SSPKRNTTSL EGNRCGNVMH ASASH | |
Greater than 90% as determined by SDS-PAGE. |
256380 | |
100 μg | |
Not determined. | |
Full Length of Isoform 3 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |