missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Serine / arginine-rich splicing factor 9 Protein
A DNA sequence encoding the Homo sapiens (Human) Serine/arginine-rich splicing factor 9, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8312-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003760.1 | |
Serine / arginine-rich splicing factor 9 Protein | |
Research Use Only | |
SFRS9 | |
Recombinant Protein | |
E. coli | |
MSGWADERGG EGDGRIYVGN LPTDVREKDL EDLFYKYGRI REIELKNRHG LVPFAFVRFE DPRDAEDAIY GRNGYDYGQC RLRVEFPRTY GGRGGWPRGG RNGPPTRRSD FRVLVSGLPP SGSWQDLKDH MREAGDVCYA DVQKDGVGMV EYLRKEDMEY ALRKLDDTKF RSHEGETSYI RVYPERSTSY GYSRSRSGSR GRDSPYQSRG SPHYFSPFRP Y | |
Greater than 90% as determined by SDS-PAGE. |
8683 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |