missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human S-phase kinase-associated protein 1
A DNA sequence encoding the Homo sapiens (Human) S-phase kinase-associated protein 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6689-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_008861.2 | |
S-phase kinase-associated protein 1 | |
Research Use Only | |
SKP1 | |
Recombinant Protein | |
E. coli | |
PSIKLQSSDG EIFEVDVEIA KQSVTIKTML EDLGMDDEGD DDPVPLPNVN AAILKKVIQW CTHHKDDPPP PEDDENKEKR TDDIPVWDQE FLKVDQGTLF ELILAANYLD IKGLLDVTCK TVANMIKGKT PEEIRKTFNI KNDFTEEEEA QVRKENQWCE EK | |
Greater than 90% as determined by SDS-PAGE. |
6500 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |