missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human S-arrestin Protein
A DNA sequence encoding the Homo sapiens (Human) S-arrestin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9381-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_000532.2 | |
S-arrestin Protein | |
Research Use Only | |
SAG | |
Recombinant Protein | |
Yeast | |
MAASGKTSKS EPNHVIFKKI SRDKSVTIYL GNRDYIDHVS QVQPVDGVVL VDPDLVKGKK VYVTLTCAFR YGQEDIDVIG LTFRRDLYFS RVQVYPPVGA ASTPTKLQES LLKKLGSNTY PFLLTFPDYL PCSVMLQPAP QDSGKSCGVD FEVKAFATDS TDAEEDKIPK KSSVRLLIRK VQHAPLEMGP QPRAEAAWQF FMSDKPLHLA VSLNKEIYFH GEPIPVTVTV TNNTEKTVKK IKAFVEQVAN VVLYSSDYYV KPVAMEEAQE KVPPNSTLTK TLTLLPLLAN NRERRGIALD GKIKHEDTNL ASSTIIKEGI DRTVLGILVS YQIKVKLTVS GFLGELTSSE VATEVPFRLM HPQPEDPAKE SYQDANLVFE EFARHNLKDA GEAEEGKRDK NDVDE | |
Greater than 90% as determined by SDS-PAGE. |
6295 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |