missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Runt-related transcription factor 3 Protein
A DNA sequence encoding the Homo sapiens (Human) Runt-related transcription factor 3, was expressed in the hosts and tags indicated.
160.00 € - 2385.00 €
Specifications
Accession Number | NP_001026850.1 |
---|---|
Gene ID (Entrez) | 864 |
Name | Runt-related transcription factor 3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16084680
|
enQuireBio™
QP6632-EC-100UG |
100 μg |
530.00 €
100µg |
Estimated Shipment: 10-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16004690
|
enQuireBio™
QP6632-EC-1MG |
1 mg |
2385.00 €
1mg |
Estimated Shipment: 10-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16094680
|
enQuireBio™
QP6632-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16014690
|
enQuireBio™
QP6632-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16024690
|
enQuireBio™
QP6632-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16034690
|
enQuireBio™
QP6632-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001026850.1 | |
Runt-related transcription factor 3 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
864 | |
Research Use Only | |
RUNX3 | |
Recombinant Protein | |
E. coli | |
MRIPVDPSTS RRFTPPSPAF PCGGGGGKMG ENSGALSAQA AVGPGGRARP EVRSMVDVLA DHAGELVRTD SPNFLCSVLP SHWRCNKTLP VAFKVVALGD VPDGTVVTVM AGNDENYSAE LRNASAVMKN QVARFNDLRF VGRSGRGKSF TLTITVFTNP TQVATYHRAI KVTVDGPREP RRHRQKLEDQ TKPFPDRFGD LERLRMRVTP STPSPRGSLS TTSHFSSQPQ TPIQGTSELN PFSDPRQFDR SFPTLPTLTE SRFPDPRMHY PGAMSAAFPY SATPSGTSIS SLSVAGMPAT SRFHHTYLPP PYPGAPQNQS GPFQANPSPY HLYYGTSSGS YQFSMVAGSS SGGDRSPTRM LASCTSSAAS VAAGNLMNPS LGGQSDGVEA DGSHSNSPTA LSTPGRMDEA VWRPY | |
Greater than 90% as determined by SDS-PAGE. |