missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ripk3 Protein
A DNA sequence encoding the Homo sapiens (Human) Ripk3, was expressed in the hosts and tags indicated.
655.00 € - 1815.00 €
Specifications
Accession Number | NP_006862.2 |
---|---|
Gene ID (Entrez) | 11035 |
Name | Ripk3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16079051
|
enQuireBio™
QP9945-BA-20UG |
20 μg |
655.00 €
20µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16069051
|
enQuireBio™
QP9945-BA-100UG |
100 μg |
1815.00 €
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16089051
|
enQuireBio™
QP9945-BA-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_006862.2 | |
Ripk3 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
11035 | |
Research Use Only | |
Ripk3 | |
Recombinant Protein | |
Baculovirus | |
MSCVKLWPSG APAPLVSIEE LENQELVGKG GFGTVFRAQH RKWGYDVAVK IVNSKAISRE VKAMASLDNE FVLRLEGVIE KVNWDQDPKP ALVTKFMENG SLSGLLQSQC PRPWPLLCRL LKEVVLGMFY LHDQNPVLLH RDLKPSNVLL DPELHVKLAD FGLSTFQGGS QSGTGSGEPG GTLGYLAPEL FVNVNRKAST ASDVYSFGIL MWAVLAGREV ELPTEPSLVY EAVCNRQNRP SLAELPQAGP ETPGLEGLKE LMQLCWSSEP KDRPSFQECL PKTDEVFQMV ENNMNAAVST VKDFLSQLRS SNRRFSIPES GQGGTEMDGF RRTIENQHSR NDVMVSEWLN KLNLEEPPSS VPKKCPSLTK RSRAQEEQVP QAWTAGTSSD SMAQPPQTPE TSTFRNQMPS PTSTGTPSPG PRGNQGAERQ GMNWSCRTPE PNPVTGRPLV NIYNCSGVQV GDNNYLTMQQ TTALPTWGLA PSGKGRGLQH PPPVGSQEGP KDPEAWSRPQ GWYNHSGK | |
Greater than 80% as determined by SDS-PAGE. |