missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Regulator of G-protein signaling 7 Protein

A DNA sequence encoding the Homo sapiens (Human) Regulator of G-protein signaling 7, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6606-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16044500

  • 2385.00 € / 1mg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001269702.1
Regulator of G-protein signaling 7 Protein
Research Use Only
RGS7
Recombinant Protein
E. coli
MAQGNNYGQT SNGVADESPN MLVYRKMEDV IARMQDEKNG IPIRTVKSFL SKIPSVFSGS DIVQWLIKNL TIEDPVEALH LGTLMAAHGY FFPISDHVLT LKDDGTFYRF QTPYFWPSNC WEPENTDYAV YLCKRTMQNK ARLELADYEA ESLARLQRAF ARKWEFIFMQ AEAQAKVDKK RDKIERKILD SQERAFWDVH RPVPGCVNTT EVDIKKSSRM RNPHKTRKSV YGLQNDIRSH SPTHTPTPET KPPTEDELQQ QIKYWQIQLD RHRLKMSKVA DSLLSYTEQY LEYDPFLLPP DPSNPWLSDD TTFWELEASK EPSQQRVKRW GFGMDEALKD PVGREQFLKF LESEFSSENL RFWLAVEDLK KRPIKEVPSR VQEIWQEFLA PGAPSAINLD SKSYDKTTQN VKEPGRYTFE DAQEHIYKLM KSDSYPRFIR SSAYQELLQA KKKSGNSMDR RTSFEKFAQN VGKSLTSKRL TSLAQSY
Greater than 90% as determined by SDS-PAGE.
6000
1 mg
Not determined.
Full Length of Isoform 5
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions