missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ras-related protein Rab-3C Protein
A DNA sequence encoding the Homo sapiens (Human) Ras-related protein Rab-3C, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7937-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001304844.1 | |
Ras-related protein Rab-3C Protein | |
Research Use Only | |
RAB3C | |
Recombinant Protein | |
E. coli | |
MRHEAPMQMA SAQDARYGQK DSSDQNFDYM FKLLIIGNSS VGKTSFLFRY ADDSFTSAFV STVGIDFKVK TVFKNEKRIK LQIWDTAGQE RYRTITTAYY RGAMGFILMY DITNEESFNA VQDWSTQIKT YSWDNAQVIL VGNKCDMEDE RVISTERGQH LGEQLGFEFF ETSAKDNINV KQTFERLVDI ICDKMSESLE TDPAITAAKQ NTRLKETPPP PQPNCAC | |
Greater than 90% as determined by SDS-PAGE. |
115827 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |