Learn More
enQuireBio™ Recombinant Human Ras association domain-containing protein 1
A DNA sequence encoding the Homo sapiens (Human) Ras association domain-containing protein 6, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP7763-ec-100ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16090651
  • 655.00 € / 100µg
Estimated Shipment: 25-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Specifications
NP_001257320.1
Ras association domain-containing protein 1
Research Use Only
RASSF6
Recombinant Protein
E. coli
MTMMAHQYPS WIFINEKTFI TREQLNSLLK TYNIFYENQK NLHILYGETE DGKLIVEGML DIFWGVKRPI QLKIQDEKPF SSFTSMKSSD VFSSKGMTRW GEFDDLYRIS ELDRTQIPMS EKRNSQEDYL SYHSNTLKPH AKDEPDSPVL YRTMSEAALV RKRMKPLMMD RKERQKNRAS INGHFYNHEI ENSPQDFALH IIFATGEQRR LKKTDIPLLQ RLLQGPSEKN ARIFLMDKDA EEISSDVAQY INFHFSLLES ILQRLNEEEK REIQRIVTKF NKEKAIILKC LQNKLVIKTE TTV
Greater than 80% as determined by SDS-PAGE.
166824
100 μg
Not determined.
Full Length of Isoform 4
Human
GST
Tris-based buffer, 50% glycerol