missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RAB10 Protein
A cDNA sequence encoding the RAB10 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13228-2ug
Additional Details : Weight : 0.01000kg
Specifications
RAB10 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAKKTYDLLF KLLLIGDSGV GKTCVLFRFS DDAFNTTFIS TIGIDFKIKT VELQGKKIKL QIWDTAGQER FHTITTSYYR GAMGIMLVYD ITNGKSFENI SKWLRNIDEH ANEDVERMLL GNKCDMDDKR VVPKGKGEQI AREHGIRFFE TSAKANINIE KAFLTLAEDI LRKTPVKEPN SENVDISSGG GVTGWKSKCC | |
Greater than 90% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The RAB10 solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol 0.4M Urea. |