missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Purine nucleoside phosphorylase / PNP Protein
A DNA sequence encoding the Homo sapiens (Human) Purine nucleoside phosphorylase / PNP, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7263-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000261.2 | |
Purine nucleoside phosphorylase / PNP Protein | |
Research Use Only | |
PNP | |
Recombinant Protein | |
E. coli | |
MENGYTYEDY KNTAEWLLSH TKHRPQVAII CGSGLGGLTD KLTQAQIFDY GEIPNFPRST VPGHAGRLVF GFLNGRACVM MQGRFHMYEG YPLWKVTFPV RVFHLLGVDT LVVTNAAGGL NPKFEVGDIM LIRDHINLPG FSGQNPLRGP NDERFGDRFP AMSDAYDRTM RQRALSTWKQ MGEQRELQEG TYVMVAGPSF ETVAECRVLQ KLGADAVGMS TVPEVIVARH CGLRVFGFSL ITNKVIMDYE SLEKANHEEV LAAGKQAAQK LEQFVSILMA SIPLPDKAS | |
Greater than 90% as determined by SDS-PAGE. |
4860 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |