missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PTPRB Protein
A DNA sequence encoding the Homo sapiens (Human) PTPRB, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6556-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001103224.1 | |
PTPRB Protein | |
Research Use Only | |
PTPRB | |
Recombinant Protein | |
Yeast | |
RQKVSHGRER PSARLSIRRD RPLSVHLNLG QKGNRKTSCP IKINQFEGHF MKLQADSNYL LSKEYEELKD VGRNQSCDIA LLPENRGKNR YNNILPYDAT RVKLSNVDDD PCSDYINASY IPGNNFRREY IVTQGPLPGT KDDFWKMVWE QNVHNIVMVT QCVEKGRVKC DHYWPADQDS LYYGDLILQM LSESVLPEWT IREFKICGEE QLDAHRLIRH FHYTVWPDHG VPETTQSLIQ FVRTVRDYIN RSPGAGPTVV HCSAGVGRTG TFIALDRILQ QLDSKDSVDI YGAVHDLRLH RVHMVQTECQ YVYLHQCVRD VLRARKLRSE QENPLFPIYE NVNPEYHRDP VYSRH | |
Greater than 90% as determined by SDS-PAGE. |
5787 | |
10 μg | |
Not determined. | |
Cytoplasmic Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |