missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PSME2 / PA28b Protein
A DNA sequence encoding the Homo sapiens (Human) PSME2 / PA28b, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8350-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_002809.2 | |
PSME2 / PA28b Protein | |
Research Use Only | |
PSME2 | |
Recombinant Protein | |
E. coli | |
AKPCGVRLSG EARKQVEVFR QNLFQEAEEF LYRFLPQKII YLNQLLQEDS LNVADLTSLR APLDIPIPDP PPKDDEMETD KQEKKEVHKC GFLPGNEKVL SLLALVKPEV WTLKEKCILV ITWIQHLIPK IEDGNDFGVA IQEKVLERVN AVKTKVEAFQ TTISKYFSER GDAVAKASKE THVMDYRALV HERDEAAYGE LRAMVLDLRA FYAELYHIIS SNLEKIVNPK GEEKPSMY | |
Greater than 90% as determined by SDS-PAGE. |
5721 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |