missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein FAM46C Protein
A DNA sequence encoding the Homo sapiens (Human) Protein FAM46C, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7730-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_060179.2 | |
Protein FAM46C Protein | |
Research Use Only | |
FAM46C | |
Recombinant Protein | |
Yeast | |
MAEESSCTRD CMSFSVLNWD QVSRLHEVLT EVVPIHGRGN FPTLEITLKD IVQTVRSRLE EAGIKVHDVR LNGSAAGHVL VKDNGLGCKD LDLIFHVALP TEAEFQLVRD VVLCSLLNFL PEGVNKLKIS PVTLKEAYVQ KLVKVCTDTD RWSLISLSNK NGKNVELKFV DSIRRQFEFS VDSFQIILDS LLFFYDCSNN PISEHFHPTV IGESMYGDFE EAFDHLQNRL IATKNPEEIR GGGLLKYSNL LVRDFRPTDQ EEIKTLERYM CSRFFIDFPD ILEQQRKLET YLQNHFAEEE RSKYDYLMIL RRVVNESTVC LMGHERRQTL NLISLLALRV LAEQNIIPSA TNVTCYYQPA PYVSDGNFSN YYVAHPPVTY SQPYPTWLPC N | |
Greater than 90% as determined by SDS-PAGE. |
54855 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |