missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Protein disulfide-isomerase TMX3 Protein

Product Code. 16021761
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16021761 50 μg 50µg
16081751 10 μg 10µg
16071751 100 μg 100µg
16001761 200 μg 200µg
16011761 500 μg 500µg
16091751 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16021761 Supplier enQuireBio™ Supplier No. QP7944ec50ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) Protein disulfide-isomerase TMX3, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_061895.3
Gene ID (Entrez) 54495
Name Protein disulfide-isomerase TMX3 Protein
Quantity 50 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol TMX3
Protein Length Full Length of Isoform 2
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His-SUMO
Sequence KGFVEDLDES FKENRNDDIW LVDFYAPWCG HCKKLEPIWN EVGLEMKSIG SPVKVGKMDA TSYSSIASEF GVRGYPTIKL LKGDLAYNYR GPRTKDDIIE FAHRVSGALI RPLPSQQMFE HMQKRHRVFF VYVGGESPLK EKYIDAASEL IVYTYFFSAS EEVVPEVIFK I
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.