missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein atonal homolog 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Protein atonal homolog 1, was expressed in the hosts and tags indicated.
160.00 € - 2385.00 €
Specifications
Accession Number | NP_005163.1 |
---|---|
Gene ID (Entrez) | 474 |
Name | Protein atonal homolog 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16091671
|
enQuireBio™
QP7932-EC-100UG |
100 μg |
530.00 €
100µg |
Estimated Shipment: Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16011681
|
enQuireBio™
QP7932-EC-1MG |
1 mg |
2385.00 €
1mg |
Estimated Shipment: Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16001681
|
enQuireBio™
QP7932-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16021681
|
enQuireBio™
QP7932-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16031681
|
enQuireBio™
QP7932-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16041681
|
enQuireBio™
QP7932-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_005163.1 | |
Protein atonal homolog 1 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
474 | |
Research Use Only | |
ATOH1 | |
Recombinant Protein | |
E. coli | |
MSRLLHAEEW AEVKELGDHH RQPQPHHLPQ PPPPPQPPAT LQAREHPVYP PELSLLDSTD PRAWLAPTLQ GICTARAAQY LLHSPELGAS EAAAPRDEVD GRGELVRRSS GGASSSKSPG PVKVREQLCK LKGGVVVDEL GCSRQRAPSS KQVNGVQKQR RLAANARERR RMHGLNHAFD QLRNVIPSFN NDKKLSKYET LQMAQIYINA LSELLQTPSG GEQPPPPPAS CKSDHHHLRT AASYEGGAGN ATAAGAQQAS GGSQRPTPPG SCRTRFSAPA SAGGYSVQLD ALHFSTFEDS ALTAMMAQKN LSPSLPGSIL QPVQEENSKT SPRSHRSDGE FSPHSHYSDS DEAS | |
Greater than 90% as determined by SDS-PAGE. |