missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Prolactin-inducible Protein

A DNA sequence encoding the Homo sapiens (Human) Prolactin-inducible protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6495-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16073720

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_002643.1
Prolactin-inducible Protein
Research Use Only
Pip
Recombinant Protein
E. coli
QDNTRKIIIK NFDIPKSVRP NDEVTAVLAV QTELKECMVV KTYLISSIPL QGAFNYKYTA CLCDDNPKTF YWDFYTNRTV QIAAVVDVIR ELGICPDDAA VIPIKNNRFY TIEILKVE
Greater than 90% as determined by SDS-PAGE.
5304
50 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions