missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Programmed cell death protein 1

A DNA sequence encoding the Homo sapiens (Human) Programmed cell death protein 5, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6475-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16043570

  • 2365.00 € / 1mg

Estimated Shipment: 16-11-2022
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_004699.1
Programmed cell death protein 1
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
9141
Greater than 80% as determined by SDS-PAGE.
Research Use Only
PDCD5
Recombinant Protein
E. coli
ADEELEALRR QRLAELQAKH GDPGDAAQQE AKHREAEMRN SILAQVLDQS ARARLSNLAL VKPEKTKAVE NYLIQMARYG QLSEKVSEQG LIEILKKVSQ QTEKTTTVKF NRRKVMDSDE DDDY
Videos
SDS
Documents
Certificates
Promotions

Promotions