missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Probable E3 SUMO-protein ligase RNF212 Protein
A DNA sequence encoding the Homo sapiens (Human) Probable E3 SUMO-protein ligase RNF212, was expressed in the hosts and tags indicated.
245.00 € - 3405.00 €
Specifications
Accession Number | NP_001124506.1 |
---|---|
Gene ID (Entrez) | 285498 |
Name | Probable E3 SUMO-protein ligase RNF212 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16084530
|
enQuireBio™
QP6612-EC-100UG |
100 μg |
770.00 €
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16004540
|
enQuireBio™
QP6612-EC-1MG |
1 mg |
3405.00 €
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16094530
|
enQuireBio™
QP6612-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16014540
|
enQuireBio™
QP6612-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16024540
|
enQuireBio™
QP6612-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16034540
|
enQuireBio™
QP6612-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001124506.1 | |
Probable E3 SUMO-protein ligase RNF212 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
285498 | |
Research Use Only | |
RNF212 | |
Recombinant Protein | |
E. coli | |
MANWVFCNRC FQPPHRTSCF SLTNCGHVYC DACLGKGKKN ECLICKAPCR TVLLSKHTDA DIQAFFMSID SLCKKYSRET SQILEFQEKH RKRLLAFYRE KISRLEESLR KSVLQIEQLQ SMRSSQQTAF STIKSSVSTK PHGCLLPPHS SAPDRLESME VDLSPSPIRK SEIAAGPARI SMISPPQDGR MGPHLTASFC FIPWLTLSKP PVPGECVISR GSPCFCIDVC PHWLLLLAFS SGRHGELTNS KTLPIYAEVQ RAVLFPFQQA EGTLDTFRTP AVSVVFPLCQ FERKKSF | |
Greater than 80% as determined by SDS-PAGE. |