Learn More
enQuireBio™ Recombinant Human PLEKHB2 Protein
A DNA sequence encoding the Homo sapiens (Human) PLEKHB2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7936-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001094093.1 | |
PLEKHB2 Protein | |
Research Use Only | |
PLEKHB2 | |
Recombinant Protein | |
E. coli | |
MAFVKSGWLL RQSTILKRWK KNWFDLWSDG HLIYYDDQTR QNIEDKVHMP MDCINIRTGQ ECRDTQPPDG KSKDCMLQIV CRDGKTISLC AESTDDCLAW KFTLQDSRTN TAYVGSAVMT DETSVVSSPP PYTAYAAPAP EQAYGYGPYG GAYPPGTQVV YAANGQAYAV PYQYPYAGLY GQQPANQVII RERYRDNDSD LALGMLAGAA TGMALGSLFW VF | |
Greater than 90% as determined by SDS-PAGE. |
55041 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |