missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PLEKHB2 Protein

A DNA sequence encoding the Homo sapiens (Human) PLEKHB2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7936-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16021711

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001094093.1
PLEKHB2 Protein
Research Use Only
PLEKHB2
Recombinant Protein
E. coli
MAFVKSGWLL RQSTILKRWK KNWFDLWSDG HLIYYDDQTR QNIEDKVHMP MDCINIRTGQ ECRDTQPPDG KSKDCMLQIV CRDGKTISLC AESTDDCLAW KFTLQDSRTN TAYVGSAVMT DETSVVSSPP PYTAYAAPAP EQAYGYGPYG GAYPPGTQVV YAANGQAYAV PYQYPYAGLY GQQPANQVII RERYRDNDSD LALGMLAGAA TGMALGSLFW VF
Greater than 90% as determined by SDS-PAGE.
55041
200 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions