missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Placental Lactogen / CSH1 Protein

A DNA sequence encoding the Homo sapiens (Human) Placental Lactogen / CSH1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8713-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16026091

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001308.1
Placental Lactogen / CSH1 Protein
Research Use Only
CSH1
Recombinant Protein
E. coli
VQTVPLSRLF DHAMLQAHRA HQLAIDTYQE FEETYIPKDQ KYSFLHDSQT SFCFSDSIPT PSNMEETQQK SNLELLRISL LLIESWLEPV RFLRSMFANN LVYDTSDSDD YHLLKDLEEG IQTLMGRLED GSRRTGQILK QTYSKFDTNS HNHDALLKNY GLLYCFRKDM DKVETFLRMV QCRSVEGSCG F
Greater than 90% as determined by SDS-PAGE.
1442
1 mg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions