missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PIGF / PLGF Protein
A DNA sequence encoding the Homo sapiens (Human) PIGF / PLGF, was expressed in the hosts and tags indicated.
160.00 € - 2385.00 €
Specifications
Accession Number | NP_001193941.1 |
---|---|
Gene ID (Entrez) | 5228 |
Name | PIGF / PLGF Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16043640
|
enQuireBio™
QP6485-EC-100UG |
100 μg |
625.00 €
100µg |
Estimated Shipment: 08-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16063640
|
enQuireBio™
QP6485-EC-1MG |
1 mg |
2385.00 €
1mg |
Estimated Shipment: 08-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16053640
|
enQuireBio™
QP6485-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16073640
|
enQuireBio™
QP6485-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16083640
|
enQuireBio™
QP6485-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16093640
|
enQuireBio™
QP6485-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001193941.1 | |
PIGF / PLGF Protein | |
Not determined. | |
Full Length of Isoform PlGF-2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
5228 | |
Research Use Only | |
PGF | |
Recombinant Protein | |
E. coli | |
LPAVPPQQWA LSAGNGSSEV EVVPFQEVWG RSYCRALERL VDVVSEYPSE VEHMFSPSCV SLLRCTGCCG DENLHCVPVE TANVTMQLLK IRSGDRPSYV ELTFSQHVRC ECRPLREKMK PERCGDAVPR R | |
Greater than 90% as determined by SDS-PAGE. |